Learn More
Research Products International Corp Cardboard Micro-Tube Storage Box Set w/ Partition, 49 Capacity
SDP
Catalog No. 501978702
Encompass
RPI's cardboard storage box is made of heavy duty cardboard and is ideal for freezer and refrigerator storage. Boxes contain a cardboard cell partition to hold popular size micro-tubes (0.2 - 2.0 ml).
Helps maximize space
Boxes have a moisture resistant covering
Catalog No. 50-197-8702 Supplier Research Products International Corp Supplier No. 181049
RPI's cardboard storage box is made of heavy duty cardboard and is ideal for freezer and refrigerator storage. Boxes contain a cardboard cell partition to hold popular size micro-tubes (0.2 - 2.0 ml).
Helps maximize space
Boxes have a moisture resistant covering