Learn More
Research Products International Corp Cardboard Micro-Tube Storage Box with Lid and Cell Divider, 3 Inch, 49 Tube Capacity
SDP
Catalog No. 50998928
Encompass
Cardboard Storage Box, 3 Inch High, with Lid and Cell Divider, 49 Tube Capacity. Partitions accept tubes with a diameter less than 16 mm.
Catalog No. 50-998-928 Supplier Research Products International Corp Supplier No. 181042
Cardboard Storage Box, 3 Inch High, with Lid and Cell Divider, 49 Tube Capacity. Partitions accept tubes with a diameter less than 16 mm.