Learn More
Andwin Scientific CARDBOARD TUBE STORAGE BOX 3IN
SDP
Catalog No. NC9749278
Encompass
The Heathrow Scientific Cardboard Cryogenic Vial Boxes are efficient products for keeping cryogenic vials in liquid nitrogen or in mechanical freezers. This product can be used with most LN2 and mechanical square or upright storage racks and can endure temperatures down to -80C. The boxes offer efficient and easy organization and identification with an imprinted label. This product also features drain holes to vent LN2 and aid in removal of condensation. It is sold in a pack of 12 boxes with included lids.
Catalog No. NC9749278 Supplier Andwin Scientific Supplier No. EF2860B
The Heathrow Scientific Cardboard Cryogenic Vial Boxes are efficient products for keeping cryogenic vials in liquid nitrogen or in mechanical freezers. This product can be used with most LN2 and mechanical square or upright storage racks and can endure temperatures down to -80C. The boxes offer efficient and easy organization and identification with an imprinted label. This product also features drain holes to vent LN2 and aid in removal of condensation. It is sold in a pack of 12 boxes with included lids.