Learn More
IMEB, Inc Tek-Select® Corrugated Cardboard, Cassette Storage Files, Assembled (2 Sets Of 5 Drawers), Durable Corrugated Cardboard, Strong Yet Lightweight, Moisture Resistant with Special Wax Coating
SDP
Catalog No. 501689492
Encompass_Preferred
Added outer corrugated shell acts as a stabilizer in keeping units stacked together over a long period of time. Each box section accommodates 2,000 microscope slides or 500 cassettes. Total capacity of each case of microscope slides is 20,000 and 5,000 cassettes. 10 drawers per case ( 2 sets of 5 drawers). Dimensions of individual box: 11 1/2 in D x 12 in W x 3 1/3 in H , Overall dimensions w/ outer shell 11 3/4 in D x 12 in W x 19 3/4 in H.
Catalog No. 50-168-9492 Supplier IMEB, Inc Supplier No. FC8A
Added outer corrugated shell acts as a stabilizer in keeping units stacked together over a long period of time. Each box section accommodates 2,000 microscope slides or 500 cassettes. Total capacity of each case of microscope slides is 20,000 and 5,000 cassettes. 10 drawers per case ( 2 sets of 5 drawers). Dimensions of individual box: 11 1/2 in D x 12 in W x 3 1/3 in H , Overall dimensions w/ outer shell 11 3/4 in D x 12 in W x 19 3/4 in H.