Learn More
IMEB, Inc Tek-Select® Corrugated Cardboard, Microscope Slides Storage Files, Unassembled (2 Sets Of 5), Durable Corrugated Cardboard that is Strong Yet Lightweight & Moisture Resistant w/Special Wax Coating
SDP
Catalog No. 501689489
Encompass_Preferred
Added outer corrugated shell acts as a stabilizer in keeping units stacked together over a long period of time. Each box section accommodates 2,000 microscope slides or 500 cassettes. Total capacity of each case of microscope slides is 20,000 and 5,000 cassettes. 10 boxes per case. Dimensions of individual box: 11 1/2 in D x 12 in W x 3 1/3 in H , Overall dimensions w/ outer shell 11 3/4 in D x 12 in W x 19 3/4 in H.
Catalog No. 50-168-9489 Supplier IMEB, Inc Supplier No. FS4
Added outer corrugated shell acts as a stabilizer in keeping units stacked together over a long period of time. Each box section accommodates 2,000 microscope slides or 500 cassettes. Total capacity of each case of microscope slides is 20,000 and 5,000 cassettes. 10 boxes per case. Dimensions of individual box: 11 1/2 in D x 12 in W x 3 1/3 in H , Overall dimensions w/ outer shell 11 3/4 in D x 12 in W x 19 3/4 in H.