Learn More
Research Products International Corp Cardboard Micro-Tube Storage Box with Lid and Cell Divider, 5 1/4'' x 5 1/4'' x 2 7/8'', 81 Tube Capacity
SDP
Catalog No. 50998926
Encompass
RPI's cardboard storage box is made of heavy duty cardboard and is ideal for freezer and refrigerator use. Boxes contain a cardboard cell partition to help hold popular size tubes. Partitions accept tubes with a diameter less than 12.70 mm.
Helps maximize space
Boxes have a moisture resistant covering
Catalog No. 50-998-926 Supplier Research Products International Corp Supplier No. 181036
RPI's cardboard storage box is made of heavy duty cardboard and is ideal for freezer and refrigerator use. Boxes contain a cardboard cell partition to help hold popular size tubes. Partitions accept tubes with a diameter less than 12.70 mm.
Helps maximize space
Boxes have a moisture resistant covering