Learn More
Sciencell Research Laboratories CARDIAC MYOCYTE MEDIUM 500ML
Catalog No. NC9548259
Encompass
Cardiac Myocyte medium is a complete medium designed for optimal growth of normal human cardiac myocytes in vitro. It is shipped as a kit with cardiac myocyte growth supplement and Pen/Strep solution.
Catalog No. NC9548259 Supplier Sciencell Research Laboratories Supplier No. 6201
Cardiac Myocyte medium is a complete medium designed for optimal growth of normal human cardiac myocytes in vitro. It is shipped as a kit with cardiac myocyte growth supplement and Pen/Strep solution.