Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZOLL™ Cardiac Science™ Semi-Recessed AED Wall Cabinet

Catalog No. 17478097 Shop All Zoll Medical Corporation Products
Change view
Click to view available options
Product Type:
Cabinet, AED
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17-478-097 Cabinet, AED
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-478-097 Supplier ZOLL™ Supplier No. 500039510

missing translation for 'validMainframeMsg'

Provides a way to store AED in accessible location, white cabinet with see-through door

  • Internal dimensions (H x W x D): 14.25 x 14.25 x 7 in.
  • External dimensions (H x W x D): 17.5 x 17.5 x 7 in.
  • Protrusion from wall: 3.12 in.
  • Included: hardware and two keys

Specifications

Product Type Cabinet, AED
Color White
Includes All hardware and two keys
Material Stainless Steel
For Use With (Equipment) ZOLL™ AEDs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.