Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cardiac Troponin I Antibody (YA3484) | 50 UL
SDP

Catalog No. 5000442687
Encompass_Preferred

Cardiac Troponin I Antibody (YA3484) is a Rabbit-derived, non-conjugated IgG monoclonal antibody targeting Cardiac Troponin I. It is supplied as a liquid and is stable for 1 year when stored at -20°C, avoiding freeze/thaw cycles. Cardiac Troponin I is the inhibitory subunit of troponin, a thin filament regulatory complex that confers calcium-sensitivity to striated muscle actomyosin ATPase activity.

  • Host: Rabbit
  • Reactivity: Human
  • Conjugation: Non-conjugated
  • SwissProt ID: P19429
  • Observed molecular weight: 28 kDa
  • Purity: Affinity purified
  • Applications: WB, IHC-P, ICC/IF, IP
  • Formulation includes glycerol for stability
  • Avoid repeated freeze/thaw cycles

Catalog No. 50-004-42687 Supplier Medchemexpress LLC Supplier No. HYP8114450UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cardiac Troponin I Antibody (YA3484) is a Rabbit-derived, non-conjugated IgG monoclonal antibody targeting Cardiac Troponin I. It is supplied as a liquid and is stable for 1 year when stored at -20°C, avoiding freeze/thaw cycles. Cardiac Troponin I is the inhibitory subunit of troponin, a thin filament regulatory complex that confers calcium-sensitivity to striated muscle actomyosin ATPase activity.

  • Host: Rabbit
  • Reactivity: Human
  • Conjugation: Non-conjugated
  • SwissProt ID: P19429
  • Observed molecular weight: 28 kDa
  • Purity: Affinity purified
  • Applications: WB, IHC-P, ICC/IF, IP
  • Formulation includes glycerol for stability
  • Avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.