Learn More
MOLECULAR DEPOT LLC Cardiac Troponin I mAb - 1 mg
Catalog No. 5000013508
Encompass_Preferred
Cardiac Troponin I Monoclonal Antibody (1 mg) is an affinity-purified mouse monoclonal antibody specific to human Cardiac Troponin I Tested in several immunoassay platforms including ELISA CLIA Immunoturbidimetry and more Several tested matching clones are available Readily available in bulk amounts and at bulk prices Biotinylation and other labeling services are also available For in vitro research use only
Catalog No. 50-000-13508 Supplier MOLECULAR DEPOT LLC Supplier No. A2010036
Cardiac Troponin I Monoclonal Antibody (1 mg) is an affinity-purified mouse monoclonal antibody specific to human Cardiac Troponin I Tested in several immunoassay platforms including ELISA CLIA Immunoturbidimetry and more Several tested matching clones are available Readily available in bulk amounts and at bulk prices Biotinylation and other labeling services are also available For in vitro research use only