Learn More
Enzo Life Sciences Cardif (human) polyclonal antibody (AT107) (100 µg)
SDP
Supplier:  Enzo Life Sciences ALX210929C100
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human Cardif (CARD adapter inducing interferon-β) (aa 160-450). UniProt: Q7Z434. Source: Purified from rabbit serum. Species Reactivity: Human. Applications: ICC, IP, WB. Recommended dilutions: Immunocytochemistry (1:500). Immunoprecipitation (1:100). Western Blot (1:2,000). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. NC0681557