Learn More
Enzo Life Sciences Cardif/MAVS (human) monoclonal antibody (58N3B6) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2590100
Encompass_Preferred
Clone: 58N3B6. Isotype: IgG2bκ. Immunogen: Human MAVS (IPS-1). Species reactivity: Human. Applications: IHC (PS), WB. Purity: Protein G purified. Formulation: Liquid. In 0.2ml PBS containing 0.05% BSA and 0.05% sodium azide.
Catalog No. 50-201-2804