Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cardinal Health™ Easy-Tie ™ Cap

Catalog No. 19160231 Shop All Cardinal Health Products
Change view
Click to view available options
Easy-Tie Cap
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-160-231 Easy-Tie Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-160-231 Supplier Cardinal Health™ Supplier No. 4359
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for particulate control

  • Adjustable ties for easy donning
  • Made of lightweight, nonwoven material

Order Info

Packaged in dispenser cartons for easy storage and access.

Specifications

Color Blue
Closure Type Ties
Packaging Type Despenser carton
For Use With (Application) Particulate control
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.