Learn More
Cardinal Health™ Limited-Use Scrub Tops
Catalog No. 191203038E Shop All Cardinal Health Products
Change view
Click to view available options
No. of Pockets:
2
Size:
Medium
Large
X-Large
2X-Large
3X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Pockets Size
19-120-3038E 2 3X-Large
19-120-3038A 2 Medium
19-120-3038B 2 Large
19-120-3038C 2 X-Large
19-120-3038D 2 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-120-3038E Supplier Cardinal Health™ Supplier No. 23506T
Add to Cart
Add to Cart
This item is not returnable. View return policy
Combines strength with the softness and comfort of linen
Unisex scrubs are constructed of high-quality, fluid-resistant, tri-layer SMS fabric. Blue.
Short Sleeve Tops
  • Feature an easy-to-don V-neck
  • Two large front pockets, one on the upper left and one on the lower righ
Specifications
Size 3X-Large
Gender Unisex
Material SMS
No. of Pockets 2
Product Type Short sleeve shirt; V-neck; Knit Cuff; Disposable