Learn More
Cardinal Health™ Low-Temperature Sterilization Pouchess
Catalog No. 19160110 Shop All Cardinal Health Products
Change view
Click to view available options
Dimensions (L x W):
25.4 x 38.1cm (10 x 15 in.)
Closure Type:
Self-seal
Catalog No. Dimensions (L x W) Closure Type
19-160-110 25.4 x 38.1cm (10 x 15 in.) Self-seal
1 options
This item is not returnable. View return policy
Catalog No. 19-160-110 Supplier Cardinal Health™ Supplier No. 32115
This item is not returnable. View return policy
Protect the sterile integrity of tools and equipment
For use with low-temperature sterilization and applications requiring EO.
  • Breathable Tyvek™ material facilitates faster aeration and easier sterilization
  • Resistant to puncture and tearing
Specifications
Closure Type Self-seal
For Use With (Application) Excellent choice for devices requiring low temperature sterilization, produced without an indicator for all types of low temperature sterilization, including EO and plasma
Material Tyvek
Dimensions (L x W) 25.4 x 38.1cm (10 x 15 in.)