Learn More
Cardinal Health™ Medi-Vac™ Guardian™ Suction Canisters
Catalog No. 19162321 Shop All Cardinal Health Products
Change view
Click to view available options
Capacity (Metric):
2 L
3 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
19-162-321 3 L
19-162-320 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-162-321 Supplier Cardinal Health™ Supplier No. 65651230
Add to Cart
Add to Cart
This item is not returnable. View return policy
With automatic overflow shutoff valve located inside lid to help prevent cross contamination of regulators and wall vacuum outlets
  • Disposable
  • 90° adapters allow tubing to connect at right angles to help prevent kinking and impeded fluid flow
  • Locking lid
  • With accessory and orthopedic ports
  • Available in a variety of sizes for almost any suction procedure
Specifications
Product Type Suction Canister
Includes Overflow shutoff valve, 90° adapters, accessory and orthopedic port
Capacity (Metric) 3 L