Learn More
Cardinal Health™ Medi-Vac™ Guardian™ Suction Canisters
Catalog No. p-4365113 Shop All Cardinal Health Products
Change view
Click to view available options
Capacity (Metric):
2 L
Catalog No. Capacity (Metric)
19-162-320 2 L
1 options
This item is not returnable. View return policy
Catalog No. 19-162-320 Supplier Cardinal Health™ Supplier No. 65651220/DEL
This item is not returnable. View return policy
With automatic overflow shutoff valve located inside lid to help prevent cross contamination of regulators and wall vacuum outlets
  • Disposable
  • 90° adapters allow tubing to connect at right angles to help prevent kinking and impeded fluid flow
  • Locking lid
  • With accessory and orthopedic ports
  • Available in a variety of sizes for almost any suction procedure
Specifications
Product Type Suction Canister
Includes Overflow shutoff valve, 90° adapters, accessory and orthopedic port
Capacity (Metric) 2 L