Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cardinal Health™ Multiple Inner Pouches

Catalog No. 19160138 Shop All Cardinal Health Products
Change view
Click to view available options
Width (Metric):
3.2 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric)
19-160-138 3.2 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-160-138 Supplier Cardinal Health™ Supplier No. 96008

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Convenient for procedures requiring multiple items

For sterilizing and storing surgery or laboratory tools and equipment.

  • Designed with 8 inner pouches side-by-side
  • Enhances aseptic technique when using more than one of the same item
  • Width per pouch: 3.2cm (1.5 in.)

Specifications

For Use With (Application) Designed for procedures requiring multiple items
Width (English) 1.5 in.
Width (Metric) 3.2 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.