Learn More
Cardinal Health™ Sleeves with Tyvek™
Catalog No. 19047787 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Case of 60
Catalog No.
19-047-787 Case of 60
1 options
This item is not returnable. View return policy
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 19-047-787 Supplier Cardinal Health™ Supplier No. 9403N
This item is not returnable. View return policy
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
For applications requiring more protection than gloves alone
Sleeves can provide barrier protection from contamination and particulates.
  • Made of Tyvek
  • Double-stitched, reinforced seams
  • Elastic at both ends
  • 18 in. long (45.7cm)
  • White