Learn More
PTS Diagnostics Accessories for CardioCheck™ Plus Analyzer
Catalog No. 21124412
Change view
Click to view available options
Type:
Printer
eGLU Test Strips
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
21-124-412 eGLU Test Strips
21-124-411 Printer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-124-412 Supplier PTS Diagnostics Supplier No. 2713
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
50 glucose test strips and MEMo Chip
Accessories for CardioCheck Plus Analyzer (cat. no. 21124410).
  • Accessory for CardioCheck Plus Analyzer (cat. no. 21124410)
  • 50 tests per vial
Specifications
Type eGLU Test Strips
For Use With (Equipment) CardioChek Plus Analyzer
Includes 50 glucose test strips and MEMo Chip