Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cardiogenol C hydrochloride | 1049741-55-0 | 99.8% | 296.75 g/mol | C13H17ClN4O2 | 10MG
SDP

Catalog No. 5000239283
Encompass_Preferred

Cardiogenol C hydrochloride is a cardiomyogenic small-molecule research compound used to induce differentiation of pluripotent stem cells into cardiomyocytes and to study cardiac development and repair. Supplied as the hydrochloride salt with high reported purity, it is provided in small laboratory pack sizes for research applications.

  • Induces cardiomyogenic differentiation in embryonic and pluripotent stem cells.
  • High reported purity (99.8%).
  • Molecular formula C13H17ClN4O2; molecular weight 296.75 g/mol.
  • Soluble in DMSO (≥59 mg/mL) and moderately soluble in water (≈2 mg/mL).
  • Provided as a solid hydrochloride salt in small pack sizes suitable for laboratory use.
  • Store solids at 4°C; solutions stable at -80°C for up to 6 months and -20°C for 1 month.

Catalog No. 50-002-39283 Supplier Medchemexpress LLC Supplier No. HY12319A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cardiogenol C hydrochloride is a cardiomyogenic small-molecule research compound used to induce differentiation of pluripotent stem cells into cardiomyocytes and to study cardiac development and repair. Supplied as the hydrochloride salt with high reported purity, it is provided in small laboratory pack sizes for research applications.

  • Induces cardiomyogenic differentiation in embryonic and pluripotent stem cells.
  • High reported purity (99.8%).
  • Molecular formula C13H17ClN4O2; molecular weight 296.75 g/mol.
  • Soluble in DMSO (≥59 mg/mL) and moderately soluble in water (≈2 mg/mL).
  • Provided as a solid hydrochloride salt in small pack sizes suitable for laboratory use.
  • Store solids at 4°C; solutions stable at -80°C for up to 6 months and -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.