Learn More
Medchemexpress LLC Cardiogenol C hydrochloride | 1049741-55-0 | 99.8% | 296.75 g/mol | C13H17ClN4O2 | 50MG
SDP
Catalog No. 5000235422
Encompass_Preferred
Cardiogenol C hydrochloride is the hydrochloride salt of a small-molecule cardiomyogenic agent used in research to induce differentiation of embryonic stem cells into cardiomyocytes. It is supplied as a solid powder and as a 10 mM solution in DMSO for in vitro biological and pharmacological studies.
Catalog No. 50-002-35422 Supplier Medchemexpress LLC Supplier No. HY12319A50MG
Cardiogenol C hydrochloride is the hydrochloride salt of a small-molecule cardiomyogenic agent used in research to induce differentiation of embryonic stem cells into cardiomyocytes. It is supplied as a solid powder and as a 10 mM solution in DMSO for in vitro biological and pharmacological studies.