Learn More
Medchemexpress LLC Cardiogenol C hydrochloride | 1049741-55-0 | MFCD06798334 | 99.8% | 296.75 g·mol⁻¹ | C13H17ClN4O2 | 100MG
SDP
Catalog No. 5000237454
Encompass_Preferred
Cardiogenol C hydrochloride is a cell-permeable pyrimidine inducer used in research to promote differentiation of embryonic stem cells into cardiomyocytes. Supplied as the hydrochloride salt, it is intended for laboratory studies requiring efficient cardiomyogenic induction and handling as a solid reagent.
  • Promotes differentiation of embryonic stem cells into cardiomyocytes.
  • Potent activity with reported EC50 ≈ 100 nM.
  • Cell-permeable small molecule suitable for in vitro studies.
  • High purity and supplied as a solid for reliable experimental use.
  • Stable when stored sealed at 4 °C, protected from moisture.
Catalog No. 50-002-37454 Supplier Medchemexpress LLC Supplier No. HY12319A100MG
Add to Cart
Add to Cart
Cardiogenol C hydrochloride is a cell-permeable pyrimidine inducer used in research to promote differentiation of embryonic stem cells into cardiomyocytes. Supplied as the hydrochloride salt, it is intended for laboratory studies requiring efficient cardiomyogenic induction and handling as a solid reagent.
  • Promotes differentiation of embryonic stem cells into cardiomyocytes.
  • Potent activity with reported EC50 ≈ 100 nM.
  • Cell-permeable small molecule suitable for in vitro studies.
  • High purity and supplied as a solid for reliable experimental use.
  • Stable when stored sealed at 4 °C, protected from moisture.