Learn More
Cayman Chemical CardIogenol ChydrochLRIde 10mg
SDP
Catalog No. 502952766
Encompass_Preferred
Induces the differentiation of MHC-positive cardiomyocytes from embryonic stem cells with an EC50 value of 0.1 µM
Catalog No. 50-295-2766 Supplier Cayman Chemical Supplier No. 1318710 MG
Induces the differentiation of MHC-positive cardiomyocytes from embryonic stem cells with an EC50 value of 0.1 µM