Learn More
Cayman Chemical Cardiogenol C (hydrochloride), Purity: ≥97%, CAS: 671225-39-1, 50 mg
SDP
Catalog No. 502952767
Encompass_Preferred
Induces the differentiation of MHC-positive cardiomyocytes from embryonic stem cells with an EC50 value of 0.1 µM
Catalog No. 50-295-2767 Supplier Cayman Chemical Supplier No. 1318750 MG
Induces the differentiation of MHC-positive cardiomyocytes from embryonic stem cells with an EC50 value of 0.1 µM