Learn More
Cardiogreen, Sigma-Aldrich
Catalog No. 01259063 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-063 25 mg
01-259-052 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-259-063 Supplier MilliporeSigma Supelco Supplier No. I263325MG
Add to Cart
Add to Cart
  • Supports protein determination by capillary electrophoresis.
  • Used with diode laser-induced fluorescence detection.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25 mg
Product Type Dye
Label Type Other Labels or Dyes
Color Green