Learn More
Medchemexpress LLC HY-D0711 500mg , Cardiogreen Foxgreen;IC Green;Indocyanine green CAS:3599-32-4 Purity:98%
SDP
Catalog No. 501492388
Encompass_Preferred
Medchemexpress, HY-D0711 500mg Cardiogreen Foxgreen;IC Green;Indocyanine green CAS:3599-32-4 Formula:C43H47N2NaO6S2 Purity:98% Cardiogreen is fluorescent agent with low toxicity has been widely used in medical diagnostics, such as determining cardiac output, hepatic function, and liver blood flow, and for ophthalmic angiography. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2388 Supplier Medchemexpress LLC Supplier No. HYD0711500MG
Medchemexpress, HY-D0711 500mg Cardiogreen Foxgreen;IC Green;Indocyanine green CAS:3599-32-4 Formula:C43H47N2NaO6S2 Purity:98% Cardiogreen is fluorescent agent with low toxicity has been widely used in medical diagnostics, such as determining cardiac output, hepatic function, and liver blood flow, and for ophthalmic angiography. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.