Learn More
Echelon Biosciences Research Labs CARDIOLIPIN BEADS 1 mL
SDP
Catalog No. 50720695
Encompass
Cardiolipin Beads are composed of agarose with 10 nanomoles of bound cardiolipin per 1ml of beads, enough for several protein binding experiments. Each 1 mL order includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-695 Supplier Echelon Biosciences Research Labs Supplier No. PBCLP1ML
Cardiolipin Beads are composed of agarose with 10 nanomoles of bound cardiolipin per 1ml of beads, enough for several protein binding experiments. Each 1 mL order includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable