Learn More
TARGETMOL CHEMICALS INC Cardiolipin Heart Bovine sodiu
SDP
Catalog No. 502876828
Encompass_Preferred
Also available in 1 mg, 5 mg and bulk. Please contact Fisher for quotes. Cardiolipin (Heart, Bovine) sodium is a phospholipid found in mitochondria and is involved in mitochondrial structure and function.Cardiolipin (Heart, Bovine) sodium is used in the study of neurological, cardiovascular, and metabolic diseases. Purity 97%
Catalog No. 50-287-6828 Supplier TARGETMOL CHEMICALS INC Supplier No. T7403110MG
Also available in 1 mg, 5 mg and bulk. Please contact Fisher for quotes. Cardiolipin (Heart, Bovine) sodium is a phospholipid found in mitochondria and is involved in mitochondrial structure and function.Cardiolipin (Heart, Bovine) sodium is used in the study of neurological, cardiovascular, and metabolic diseases. Purity 97%