Learn More
Cayman Chemical Cardiolipin (18:1/18:1/18:1/18:1) (sodium salt), Purity: ≥95%, CAS: 115404-77-8, 10 mg
SDP
Catalog No. 502955564
Encompass_Preferred
A phospholipid; has been used in the generation of LNPs; LNPs containing cardiolipin (18:1/18:1/18:1/18:1), 4A3-SCB, DMG-PEG(2000), 1,2-DOPE, and cholesterol and encapsulating mRNA accumulate in the spleen in mice
Catalog No. 50-295-5564 Supplier Cayman Chemical Supplier No. 1404510 MG
A phospholipid; has been used in the generation of LNPs; LNPs containing cardiolipin (18:1/18:1/18:1/18:1), 4A3-SCB, DMG-PEG(2000), 1,2-DOPE, and cholesterol and encapsulating mRNA accumulate in the spleen in mice