Learn More
3B Scientific™ Cardionics PAT Basic™ Pediatric Auscultation Trainer without WiFi

Shop All 3B Scientific Products


Description
- The PAT Basic™ trainer engages the technology of the SimScope stethoscope to simulate pediatric heart, lung, and bowel sounds utilizing a large sounds library
- Using the provided SimScope stethoscope, users are able to access a large variety of pathological conditions in order to customize scenarios
- By placing the SimScope over the anatomically correct locations of PAT Basic™, the programmed sounds will be played through the scope, allowing for students to employ and enhance their diagnostic skills
- PAT Basic™ consists of a Male Manikin, a SimScope, SimScope Software, Site License, USB Cable, SAM T-Shirt, and User's Manual
Specifications
Specifications
| Product Type | Manikin |
| For Use With (Application) | Teaching or Learning Pediatric Heart, Lung, and Bowel Sounds |
| Color | Light Skin |
| Includes | Male Manikin, a SimScope, SimScope Software, Site License, USB Cable, SAM T-Shirt, and User's Manual |
| Weight (English) | 9.91 lb. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.