Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Cardionics PAT Basic™ Pediatric Auscultation Trainer without WiFi
GREENER_CHOICESDPSCI_ED

Catalog No. S27947 Shop All 3B Scientific Products
Change view
Click to view available options
Color:
Dark Skin
Light Skin
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S27947 Dark Skin
S27946 Light Skin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27947 Supplier 3B Scientific™ Supplier No. 1022471
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3B Scientific™ Cardionics PAT Basic™ Pediatric Auscultation Trainer without WiFi uses a SimScope stethoscope to simulate pediatric heart, lung, and bowel sounds

  • The PAT Basic™ trainer engages the technology of the SimScope stethoscope to simulate pediatric heart, lung, and bowel sounds utilizing a large sounds library
  • Using the provided SimScope stethoscope, users are able to access a large variety of pathological conditions in order to customize scenarios
  • By placing the SimScope over the anatomically correct locations of PAT Basic™, the programmed sounds will be played through the scope, allowing for students to employ and enhance their diagnostic skills
  • PAT Basic™ consists of a Male Manikin, a SimScope, SimScope Software, Site License, USB Cable, SAM T-Shirt, and User's Manual

Specifications

Product Type Manikin
For Use With (Application) Teaching or Learning Pediatric Heart, Lung, and Bowel Sounds
Color Dark Skin
Includes Male Manikin, a SimScope, SimScope Software, Site License, USB Cable, SAM T-Shirt, and User's Manual
Weight (English) 9.91 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.