Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Cardionics SAM Basic™ Student Auscultation Manikin
GREENER_CHOICESDPSCI_ED

Catalog No. S27944 Shop All 3B Scientific Products
Change view
Click to view available options
Color:
Dark Skin
Light Skin
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S27944 Light Skin
S27945 Dark Skin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27944 Supplier 3B Scientific™ Supplier No. 1020097
Add to Cart
Add to Cart

3B Scientific™ Cardionics SAM Basic™ Student Auscultation Manikin used to teach or learn heart, lung, and bowel sounds

Product Features:

  • Auscultation is an essential clinical skill needed to assess and monitor patients' conditions
  • SAM Basic™ Trainer can be used with the SimScope stethoscope (included)
  • All sounds and videos are recorded from adult live patients for students to experience a life-like simulation
  • SimScope stethoscope is used to simulate adult heart, lung and bowel sounds, utilizing a large sound library incorporated into the SimScope stethoscope
  • By placing the SimScope over the anatomically correct locations of SAM Basic, the programmed sounds will be played through the scope, allowing for students to employ and enhance their diagnostic skills.
  • Uses include: individual learning, group lectures, and for viewing phonocardiograms

Delivery content:

  • A portable SAM Basic Male Auscultation Torso
  • One SimScope stethoscope
  • The SimScope software and site license
  • USB cable
  • One SAM t-shirt
  • User's manual

How is SAM Basic Student Auscultation Manikin different?

  • It is lightweight & portable
  • It utilizes the extensive Cardionics proprietary heart, lung, bowel and bruit sounds library
  • Students can listen at anatomically correct auscultation sites
  • Customized Library is available to fit specific scenarios and programs
  • The SimScope WiFi upgrade allows for remote selection of sound scenarios

Specifications

Product Type Manikin
For Use With (Application) Teaching or Learning Heart, Lung, and Bowel Sounds
Color Light Skin
Includes Basic Male Auscultation Torso, SimScope Stethoscope, SimScope Software, Site License, USB Cable, SAM T-Shirt, User's Manual
Weight (English) 14.55 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.