Learn More
Medchemexpress LLC Cardiotoxin analog (CTX) IV (6-12) | 115722-23-1 | 99.95% | 899.13 | 1 MG
SDP
Catalog No. 5000424197
Encompass_Preferred
Cardiotoxin analog (CTX) IV (6-12) is a membrane-active peptide derived from Taiwan cobra venom. It specifically targets negatively charged phospholipid membranes like phosphatidylserine and phosphatidylinositol. By binding to cell membranes and embedding into lipid bilayers through hydrophobic and electrostatic interactions, it destroys membrane stability.
  • Targets negatively charged phospholipid membranes
  • Isolated from Taiwan cobra venom
  • Binds to cell membranes and embeds into lipid bilayers
  • Induces membrane lipid disorder and cell lysis
  • Exhibits hemolysis and cytotoxicity
  • For research use only
Catalog No. 50-004-24197 Supplier Medchemexpress LLC Supplier No. HYP19021MG
Cardiotoxin analog (CTX) IV (6-12) is a membrane-active peptide derived from Taiwan cobra venom. It specifically targets negatively charged phospholipid membranes like phosphatidylserine and phosphatidylinositol. By binding to cell membranes and embedding into lipid bilayers through hydrophobic and electrostatic interactions, it destroys membrane stability.
  • Targets negatively charged phospholipid membranes
  • Isolated from Taiwan cobra venom
  • Binds to cell membranes and embeds into lipid bilayers
  • Induces membrane lipid disorder and cell lysis
  • Exhibits hemolysis and cytotoxicity
  • For research use only