Learn More
Cayman Chemical Cardnol dIen 10mg
SDP
Catalog No. 502987257
Encompass_Preferred
A tyrosinase inhibitor (IC50 = 52.5 μM); used to synthesize cardanol-metal complexes
Catalog No. 50-298-7257 Supplier Cayman Chemical Supplier No. 2315310 MG
A tyrosinase inhibitor (IC50 = 52.5 μM); used to synthesize cardanol-metal complexes