Learn More
Cayman Chemical Cardnol trIen 10mg
SDP
Catalog No. 502987524
Encompass_Preferred
A tyrosinase inhibitor (IC50 = 40.5 µM); found in cashew nut shell liquid; used as a mixture with cardanol di- and triene to synthesize cardanol-metal complexes that inhibit biofilm formation
Catalog No. 50-298-7524 Supplier Cayman Chemical Supplier No. 2315510 MG
A tyrosinase inhibitor (IC50 = 40.5 µM); found in cashew nut shell liquid; used as a mixture with cardanol di- and triene to synthesize cardanol-metal complexes that inhibit biofilm formation