Learn More
Cayman Chemical Cardol triene, Purity: ≥95%, CAS: 79473-24-8, 10 mg
SDP
Catalog No. 502993776
Encompass_Preferred
A phenol from cashew nut shell liquid; inhibits mushroom tyrosinase (IC50 = 2.2 µM); kills 25, 75, and 100% of S. mansoni worms after 24 hours when used at concentrations of 50, 100, or 200 µM, respectively; has been used as a starting material for the synthesis of bis-benzoxazines
Catalog No. 50-299-3776 Supplier Cayman Chemical Supplier No. 2337410 MG
A phenol from cashew nut shell liquid; inhibits mushroom tyrosinase (IC50 = 2.2 µM); kills 25, 75, and 100% of S. mansoni worms after 24 hours when used at concentrations of 50, 100, or 200 µM, respectively; has been used as a starting material for the synthesis of bis-benzoxazines