Learn More
Electron Microscopy Sciences Cardolite NC-513 225 ML
SDP
Catalog No. 50980288
Encompass_Preferred
(15-M-Phenylglycidyl-(n-pentadecca-8,11-dienyl))
CAS #171263-25-5 CH₂OCHCH₂OC₆H₄C₁₅H₂₇ F.W. 356
A Long-chain, monofunctional epoxide; stable when mixed with resins. Viscosity 50 cps at 25 degree C. Cardolite is a registered trademark of Cardolite Corporation.
Catalog No. 50-980-288 Supplier Electron Microscopy Sciences Supplier No. 12400
(15-M-Phenylglycidyl-(n-pentadecca-8,11-dienyl))
CAS #171263-25-5 CH₂OCHCH₂OC₆H₄C₁₅H₂₇ F.W. 356
A Long-chain, monofunctional epoxide; stable when mixed with resins. Viscosity 50 cps at 25 degree C. Cardolite is a registered trademark of Cardolite Corporation.