Learn More
Sigma Aldrich Fine Chemicals Biosciences Cardolpn sodum salt fr100MG
Catalog No. 502905461
Encompass
Cardiolipin is localized in the inner membrane of mitochondria. It contains 18 carbon atoms 3-phosphatidyl groups connected via a glycerol bridge. Cardiolipin is an important component of ATP synthase enzyme.
Catalog No. 50-290-5461 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C0563100MG
Cardiolipin is localized in the inner membrane of mitochondria. It contains 18 carbon atoms 3-phosphatidyl groups connected via a glycerol bridge. Cardiolipin is an important component of ATP synthase enzyme.