Learn More
MSC 2XL Multi-Surface Cleaning Wipes, ESD/Antistatic, Pre-Saturated, 12 x 8 in
Catalog No. 502839817
Encompass
These industrial strength pre-moistened towelettes effectively remove grease, grime, adhesives, dirt, and oily messes. They are versatile for cleaning hands, tools, equipment, and hard surfaces. Featuring built-in scrubbing fibers, a waterless formula that prevents rust and leaves no residue, and gentle on hands. Made from photo-degradable materials.
  • Industrial strength pre-moistened towelettes
  • Removes grease, grime, adhesives, dirt, oily messes
  • Versatile for hands, tools, equipment, and hard surfaces
  • Built-in scrubbing fibers
  • Gentle on hands; waterless, no residue, prevents rust
  • Photo-degradable materials
  • ESD/Antistatic
  • Alcohol-free
  • Multi-surface flat sheet, roll packaged
  • Meltblown construction
  • 12 in x 8 in sheet dimensions
  • Industrial and automotive use
Catalog No. 50-283-9817 Supplier MSC Supplier No. 92688092
These industrial strength pre-moistened towelettes effectively remove grease, grime, adhesives, dirt, and oily messes. They are versatile for cleaning hands, tools, equipment, and hard surfaces. Featuring built-in scrubbing fibers, a waterless formula that prevents rust and leaves no residue, and gentle on hands. Made from photo-degradable materials.
  • Industrial strength pre-moistened towelettes
  • Removes grease, grime, adhesives, dirt, oily messes
  • Versatile for hands, tools, equipment, and hard surfaces
  • Built-in scrubbing fibers
  • Gentle on hands; waterless, no residue, prevents rust
  • Photo-degradable materials
  • ESD/Antistatic
  • Alcohol-free
  • Multi-surface flat sheet, roll packaged
  • Meltblown construction
  • 12 in x 8 in sheet dimensions
  • Industrial and automotive use