Learn More
Medchemexpress LLC HY-121356 1mg Medchemexpress, Carebastine CAS:90729-42-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1213561MG
Encompass_Preferred
Medchemexpress, HY-121356 1mg Carebastine CAS:90729-42-3 Carebastine is the active metabolite of Ebastine. Carebastine is a histamine H1 receptor antagonist. Carebastine inhibits VEGF-induced HUVEC and HPAEC proliferation, migration and angiogenesis in a dose-dependent manner [1] . Carebastine suppresses the expression of macrophage migration inhibitory factor [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2557