Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ CarePacs™ 3rd Weight,2G, ASTM Class 1, C, WeightLink™

Catalog No. 01914363 Shop All METTLER TOLEDO Products
Change view
Click to view available options
CarePacs 3rd Weight,2G, ASTM, 1, C, WeightLink
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-914-363 CarePacs 3rd Weight,2G, ASTM, 1, C, WeightLink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-914-363 Supplier Mettler Toledo™ Supplier No. 30293488

missing translation for 'validMainframeMsg'

CarePacs ASTM for WeightLink. Full traceability: to ensure maximum safety and traceability in routine testing, the two weights are linked to their certificate through a unique machine readable serial number. A card containing the certificate values in the form of a data matrix code can be read with any DMC scanning device for digital data transmission.

  • Box: Plastic box (included)
  • Calibration Certificate: Yes
  • Density p: 7950 (± 140) kg/m3
  • Design: One-piece (monobloc)
  • Material: Stainless Steel
  • Susceptibility X: 1 ≤ 0.03
  • Digital Readable Certificate - A card containing the certificate values in the form of a data matrix code can be read with any DMC scanning device for digital data transmission.
TRUSTED_SUSTAINABILITY

Specifications

Product Type ASTM Weight
Classification ASTM Class 1
Material Stainless Steel
Mass 2 g
For Use With (Application) Balance calibration and testing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.