Learn More
Enterprise Technology Solutions CareTek Biohazard Waste Bag, Peel/Stick, 1.4 Qt, 9"x10", 100/BX, RD
SDP
Catalog No. 5000790747
Encompass_Preferred
These 2 mil infection-control waste bags stick up virtually anywhere with peel-and-stick adhesive strips. Just peel off protective liner and adhere bag to most any clean, dry surface. When bags are full, just peel second liner off the front, seal, and dispose of properly. Bags remove without leaving any gummy residue. Each bag displays bilingual hazard warning.
Catalog No. 50-007-90747 Supplier Enterprise Technology Solutions Supplier No. CTKCTRB042910
These 2 mil infection-control waste bags stick up virtually anywhere with peel-and-stick adhesive strips. Just peel off protective liner and adhere bag to most any clean, dry surface. When bags are full, just peel second liner off the front, seal, and dispose of properly. Bags remove without leaving any gummy residue. Each bag displays bilingual hazard warning.