Learn More
Biocare Medical, LLC Carezyme I: Trypsin Kit - 25 ml
SDP
Catalog No. 5083292
Encompass_Preferred
Trypsin is a commonly used digestive enzyme. In formalin-fixed paraffin-embedded tissues, certain antibody protocols required enzyme pretreatment for proper immunohistochemical staining. This product is provided as a two component, ready-to-use solution, and can be used at different concentrations.
Catalog No. 50-832-92 Supplier Biocare Medical, LLC Supplier No. TRP955KH
Trypsin is a commonly used digestive enzyme. In formalin-fixed paraffin-embedded tissues, certain antibody protocols required enzyme pretreatment for proper immunohistochemical staining. This product is provided as a two component, ready-to-use solution, and can be used at different concentrations.