Learn More
Biocare Medical, LLC Carezyme II: Pepsin - 25 ml
SDP
Catalog No. 5082990
Encompass_Preferred
Pepsin is a commonly used digestive enzyme. For formalin-fixed paraffin-embedded tissues, certain antibody protocols require enzyme pretreatment for proper immuno-histochemical staining. This product is provided as a single-component, ready-to-use solution. It can also be used on an automated staining system.
Catalog No. 50-829-90 Supplier Biocare Medical, LLC Supplier No. PEP956H
Pepsin is a commonly used digestive enzyme. For formalin-fixed paraffin-embedded tissues, certain antibody protocols require enzyme pretreatment for proper immuno-histochemical staining. This product is provided as a single-component, ready-to-use solution. It can also be used on an automated staining system.