Learn More
Medchemexpress LLC HY-10455 100mg , Carfilzomib CAS:868540-17-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10455100MG
Encompass_Preferred
HY-10455 100mg Carfilzomib CAS: 868540-17-4 Carfilzomib (PR-171) is an irreversible proteasome inhibitor with an IC50 of 5 nM in ANBL-6 and RPMI 8226 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9304