Learn More
Medchemexpress LLC HY-10455 10mg Medchemexpress, Carfilzomib CAS:868540-17-4 Purity:>98%
SDP
Catalog No. 502028756
Encompass_Preferred
Medchemexpress, HY-10455 10mg Carfilzomib CAS:868540-17-4 Carfilzomib is an irreversible proteasome inhibitor with an IC50 of 5 nM in ANBL-6 and RPMI 8226 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8756 Supplier Medchemexpress LLC Supplier No. HY1045510MG
Medchemexpress, HY-10455 10mg Carfilzomib CAS:868540-17-4 Carfilzomib is an irreversible proteasome inhibitor with an IC50 of 5 nM in ANBL-6 and RPMI 8226 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.