Learn More
Medchemexpress LLC HY-10455 200mg , Carfilzomib PR-171 CAS:868540-17-4 Purity:98%
SDP
Catalog No. 501492526
Encompass_Preferred
Medchemexpress, HY-10455 200mg Carfilzomib PR-171 CAS:868540-17-4 Formula:C40H57N5O7 IC50: 5 nM (Proteasome) Purity:98% Carfilzomib is an irreversible proteasome inhibitor with IC 50 of < 5 nM in ANBL-6 and RPMI 8226 cells. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2526 Supplier Medchemexpress LLC Supplier No. HY10455200MG
Medchemexpress, HY-10455 200mg Carfilzomib PR-171 CAS:868540-17-4 Formula:C40H57N5O7 IC50: 5 nM (Proteasome) Purity:98% Carfilzomib is an irreversible proteasome inhibitor with IC 50 of < 5 nM in ANBL-6 and RPMI 8226 cells. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.