Learn More
Apexbio Technology LLC Carfilzomib (PR-171) 868540-17-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063486
Encompass_Preferred
Carfilzomib (PR-171 CAS 868540-17-4) is a derivative of the natural product epoxomicin and functions as an irreversible inhibitor of the chymotrypsin-like activity within the 20S proteasome Proteasomes play a critical role in intracellular protein degradation thus inhibition results in accumulation of polyubiquitinated proteins Through this mechanism Carfilzomib induces cell cycle arrest and apoptosis consequently suppressing tumor cell growth Research applications primarily involve exploration of its antitumor efficacy and mechanisms in oncology models
Catalog No. 50-000-63486 Supplier Apexbio Technology LLC Supplier No. A19335.1
Carfilzomib (PR-171 CAS 868540-17-4) is a derivative of the natural product epoxomicin and functions as an irreversible inhibitor of the chymotrypsin-like activity within the 20S proteasome Proteasomes play a critical role in intracellular protein degradation thus inhibition results in accumulation of polyubiquitinated proteins Through this mechanism Carfilzomib induces cell cycle arrest and apoptosis consequently suppressing tumor cell growth Research applications primarily involve exploration of its antitumor efficacy and mechanisms in oncology models