Learn More
Abcam Fluorescein-5(6)-carbo x amidocaproic acid N-hydro x ysuccinimidyl ester, 5MG
Catalog No. 50996185
Encompass_Preferred
MW 586.55 g/mol, Purity >85%. Green fluorescent dye for the labeling of primary amines. Fluorescein labeling reagent with a hexanoic spacer that improves the reactive group accessibility for nucleophiles in preparation of bioconjugates. Reactive N-hydroxysuccinimide (NHS) group can be used for easy attachment to proteins or amine containing nucleotides. Excitation maximum = 491 nm; emission maximum = 515 nm .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-185 Supplier Abcam Supplier No. AB2738945MG
MW 586.55 g/mol, Purity >85%. Green fluorescent dye for the labeling of primary amines. Fluorescein labeling reagent with a hexanoic spacer that improves the reactive group accessibility for nucleophiles in preparation of bioconjugates. Reactive N-hydroxysuccinimide (NHS) group can be used for easy attachment to proteins or amine containing nucleotides. Excitation maximum = 491 nm; emission maximum = 515 nm .
The product is subject to the following: Abcam Restricted Use Statement